Please login to your online account to display your discounted pricing

anti-Caspase-7, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056115SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056115T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 840
Format Antiserum
Antigen Caspase-7
Formulation Neat serum.
Test Specificity This polyclonal antisera recognizes the large (∽17-22 kDa) and small (∽10 kDa) subunits of active/cleaved caspase-7. Whereas the antisera has a strong preference for active/cleaved caspase-7, in some cell or tissue systems or techniques the antisera may also recognize the proform of Caspase-7 (∽35 kDa). Cross reacts with Gerbil, Human, Mouse and Rat.
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Gerbil
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Conjugate Unlabeled
Quantity 0.025mL
Immunogen Recombinant catalytically active human Caspase-7 protein was used as immunogen.
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit