Please login to your online account to display your discounted pricing

anti-Caspase-7, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056115

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056115

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 840
Conjugate Unlabeled
Cross Reactivity Gerbil
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen Caspase-7
Formulation Neat serum.
Immunogen Recombinant catalytically active human Caspase-7 protein was used as immunogen.
Test Specificity This polyclonal antisera recognizes the large (∽17-22 kDa) and small (∽10 kDa) subunits of active/cleaved caspase-7. Whereas the antisera has a strong preference for active/cleaved caspase-7, in some cell or tissue systems or techniques the antisera may also recognize the proform of Caspase-7 (∽35 kDa). Cross reacts with Gerbil, Human, Mouse and Rat.
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Quantity 0.05mL
Primary or Secondary Primary
Regulatory Status RUO
Host Species Rabbit