Please login to your online account to display your discounted pricing

anti-Caspase-7, Polyclonal, Novus Biologicals™

$144.63 - $252.00

Specifications

Host Species Rabbit
Immunogen Recombinant catalytically active human Caspase-7 protein was used as immunogen.
Antigen Caspase-7
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056115

 
novus biologicals canada
NB10056115
Unlabeled 0.05mL Each for $252.00

NB10056115T

 
novus biologicals canada
NB10056115SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Recombinant catalytically active human Caspase-7 protein was used as immunogen.
Antigen Caspase-7
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity This polyclonal antisera recognizes the large (∽17-22 kDa) and small (∽10 kDa) subunits of active/cleaved caspase-7. Whereas the antisera has a strong preference for active/cleaved caspase-7, in some cell or tissue systems or techniques the antisera may also recognize the proform of Caspase-7 (∽35 kDa). Cross reacts with Gerbil, Human, Mouse and Rat.
Gene ID (Entrez) 840
Formulation Neat serum.