Please login to your online account to display your discounted pricing

anti-Caspase-3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056112

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056112

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Gene Accession No. P42574
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Gene ID (Entrez) 836
Test Specificity Caspase antibodies are classical tools for detecting inactive (pro) and active (cleaved) forms of the enzymes. The presence of the large or small subunits in western blots is considered to be a marker of caspase activation. Cross reacts with Gerbil, Human, Mouse and Rat.
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen Caspase-3
Immunogen Recombinant full-length human Caspase-3 protein (pro-form) was used as immunogen.
Host Species Rabbit
Format Antiserum
Quantity 0.05mL