Please login to your online account to display your discounted pricing

anti-Caspase-3, Polyclonal, Novus Biologicals™

$144.63 - $144.63

Specifications

Host Species Rabbit
Immunogen Recombinant full-length human Caspase-3 protein (pro-form) was used as immunogen.
Antigen Caspase-3
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056112T

 
novus biologicals canada
NB10056112SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Recombinant full-length human Caspase-3 protein (pro-form) was used as immunogen.
Antigen Caspase-3
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blot
Test Specificity Caspase antibodies are classical tools for detecting inactive (pro) and active (cleaved) forms of the enzymes. The presence of the large or small subunits in western blots is considered to be a marker of caspase activation. Cross reacts with Gerbil, Human, Mouse and Rat.
Gene Accession No. P42574
Gene ID (Entrez) 836
Formulation PBS