Please login to your online account to display your discounted pricing

anti-Caspase-12, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224618SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224618SS

  • / Each
 

Description & Specifications

Specifications

Antigen Caspase-12
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Gene Accession No. Q6UXS9
Host Species Rabbit
Immunogen Made to amino acids between 100 and 150 of human Caspase-12.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 100506742
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, 94% homologous in monkey, 88% homologous in cow and dog, and 83% homologous in mouse and rat.
Target Species Human