Please login to your online account to display your discounted pricing

anti-Caspase-12, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224618

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224618

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Quantity 0.1mg
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, 94% homologous in monkey, 88% homologous in cow and dog, and 83% homologous in mouse and rat.
Concentration 0.5mg/mL
Gene Accession No. Q6UXS9
Gene ID (Entrez) 100506742
Immunogen Made to amino acids between 100 and 150 of human Caspase-12.
Antigen Caspase-12
Primary or Secondary Primary
Conjugate Unlabeled
Applications Western Blotting
Host Species Rabbit
Cross Reactivity Human
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Format Purified