Please login to your online account to display your discounted pricing

anti-Caspase-12, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen Made to amino acids between 100 and 150 of human Caspase-12.
Antigen Caspase-12
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224618

 
novus biologicals canada
NBP224618
Unlabeled 0.1mg Each for $257.41

NBP224618SS

 
novus biologicals canada
NBP224618SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Made to amino acids between 100 and 150 of human Caspase-12.
Antigen Caspase-12
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, 94% homologous in monkey, 88% homologous in cow and dog, and 83% homologous in mouse and rat.
Gene Accession No. Q6UXS9
Gene ID (Entrez) 100506742
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL