Please login to your online account to display your discounted pricing

anti-Caspase 10 Polyclonal, Thermo Scientific™

Caspase 10 Polyclonal antibody PA1-12507 from Thermo Scientific™ specifically detects Caspase 10 in Human, Mouse and Rat samples, and it is validated for Western Blot

$448.05 - $448.05

Specifications

Antigen Caspase 10
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-674-167

 
pierce biotechnology inc
PA112507
Unlabeled 100μg Each for $448.05
Description & Specifications

Specifications

Antigen Caspase 10
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues 505-521 of human FLICE2.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

Caspase 10 Polyclonal antibody PA1-12507 from Thermo Scientific™ specifically detects Caspase 10 in Human, Mouse and Rat samples, and it is validated for Western Blot

Caspases are cysteine proteases, expressed as inactive precursors, that mediate apoptosis by proteolysis of specific substrates. Caspases have the ability to cleave after aspartic acid residues. There are two classes of caspases involved in apoptosis; initiators (activation by receptor cluster) and effectors (activation by mitochondrial permeability transition). Proapoptotic signals autocatalytically activate initiator caspases, such as Caspase-8 and Caspase 9. Activated initiator caspases then process effector caspases, such as Caspase-3 and Caspase-7, which in turn cause cell collapse.
Host Species: Rabbit
Cross Reactivity: Human, Mouse, Rat
Isotype: IgG
Immunogen: Synthetic peptide corresponding to residues 505-521 of human FLICE2.
Formulation: 100μg purified Antibody in PBS, with 0.02% sodium azide;
Synonyms: ALPS2, FLICE2, MCH4
Uniprot Accession: Q92851
Gene Symbol: CASP10
Entrez GeneID: 843
PA1-12507 detects Caspase 10 in Human, Mouse and Rat samples. PA1-12507 has been successfully used in Western Blot procedures. PA1-12507 The immunogen is Synthetic peptide corresponding to residues 505-521 of human FLICE2..