Please login to your online account to display your discounted pricing

anti-Caspase-10/FLICE2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056123

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056123

  • / Each
 

Description & Specifications

Specifications

Antigen Caspase-10/FLICE2
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Antisera
Formulation PBS
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminal aa 435-455 (QPAFTLRKKLVFPVPLDALSI
Quantity 0.05mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 843
Test Specificity This polyclonal antisera recognizes caspase-10 forms that contain the peptide immunogen sequence (QPAFTLRKKLVFPVPLDALSI). Cross reacts with Dog, Gerbil, Human, Mouse and Rat.
Target Species Canine
Target Species Human
Target Species Murine
Target Species Rat