Please login to your online account to display your discounted pricing

anti-Caspase-10/FLICE2, Polyclonal, Novus Biologicals™

$144.63 - $222.95

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to aa 373-392 (ALNPEQAPTSLQDSIPAEAD) of human caspase 10a was used as immunogen
Antigen Caspase-10/FLICE2
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056124

 
novus biologicals canada
NB10056124
Unlabeled 0.05mL Each for $222.95

NB10056124T

 
novus biologicals canada
NB10056124SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to aa 373-392 (ALNPEQAPTSLQDSIPAEAD) of human caspase 10a was used as immunogen
Antigen Caspase-10/FLICE2
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity This polyclonal antisera recognizes caspase-10 forms that contain the peptide immunogen sequence, amino acids 373-392 (ALNPEQAPTSLQDSIPAEAD) of human caspase 10, including caspase-10a, caspase-10b, and caspase-10d.
Gene ID (Entrez) 843
Formulation PBS