Please login to your online account to display your discounted pricing

anti-Caspase-10/FLICE2, Polyclonal, Novus Biologicals™

$144.63 - $222.95

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminal aa 435-455 (QPAFTLRKKLVFPVPLDALSI
Antigen Caspase-10/FLICE2
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056123

 
novus biologicals canada
NB10056123
Unlabeled 0.05mL Each for $222.95

NB10056123T

 
novus biologicals canada
NB10056123SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminal aa 435-455 (QPAFTLRKKLVFPVPLDALSI
Antigen Caspase-10/FLICE2
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity This polyclonal antisera recognizes caspase-10 forms that contain the peptide immunogen sequence (QPAFTLRKKLVFPVPLDALSI). Cross reacts with Dog, Gerbil, Human, Mouse and Rat.
Gene ID (Entrez) 843
Formulation PBS