Please login to your online account to display your discounted pricing

anti-Caspase-10/FLICE2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056124SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056124T

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 843
Test Specificity This polyclonal antisera recognizes caspase-10 forms that contain the peptide immunogen sequence, amino acids 373-392 (ALNPEQAPTSLQDSIPAEAD) of human caspase 10, including caspase-10a, caspase-10b, and caspase-10d.
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Host Species Rabbit
Immunogen A synthetic peptide corresponding to aa 373-392 (ALNPEQAPTSLQDSIPAEAD) of human caspase 10a was used as immunogen
Quantity 0.025mL
Antigen Caspase-10/FLICE2
Conjugate Unlabeled
Cross Reactivity Human
Regulatory Status RUO
Format Antiserum