Please login to your online account to display your discounted pricing

anti-Caspase-10/FLICE2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056124

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056124

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Primary or Secondary Primary
Gene ID (Entrez) 843
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity This polyclonal antisera recognizes caspase-10 forms that contain the peptide immunogen sequence, amino acids 373-392 (ALNPEQAPTSLQDSIPAEAD) of human caspase 10, including caspase-10a, caspase-10b, and caspase-10d.
Regulatory Status RUO
Format Antiserum
Immunogen A synthetic peptide corresponding to aa 373-392 (ALNPEQAPTSLQDSIPAEAD) of human caspase 10a was used as immunogen
Host Species Rabbit
Antigen Caspase-10/FLICE2
Cross Reactivity Human
Quantity 0.05mL