Please login to your online account to display your discounted pricing

anti-Caspase-10/FLICE2, Polyclonal, Novus Biologicals™

Manufacturer: novus biologicals canada  NB10056123SS

 View all options for this product


For Research Use Only

Catalog No. NB10056123T


Description & Specifications


Antigen Caspase-10/FLICE2
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Antiserum
Formulation PBS
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminal aa 435-455 (QPAFTLRKKLVFPVPLDALSI
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 843
Test Specificity This polyclonal antisera recognizes caspase-10 forms that contain the peptide immunogen sequence (QPAFTLRKKLVFPVPLDALSI). Cross reacts with Dog, Gerbil, Human, Mouse and Rat.