Please login to your online account to display your discounted pricing

anti-Carm1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056374

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056374

  • / Each
 

Description & Specifications

Specifications

Formulation PBS containing 0.05% BSA
Primary or Secondary Primary
Concentration 0.5mg/mL
Cross Reactivity Human
Conjugate Unlabeled
Gene Accession No. Q86X55
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 10498
Regulatory Status RUO
Applications Western Blotting
Antigen Carm1
Immunogen This antibody was developed against a mixture of synthetic peptides containing amino acids 45-69 and 595-608 of human PRMT4.
Format Purified
Host Species Rabbit
Quantity 0.1mg