Please login to your online account to display your discounted pricing

anti-Carm1, Polyclonal, Novus Biologicals™

$139.00 - $257.41

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a mixture of synthetic peptides containing amino acids 45-69 and 595-608 of human PRMT4.
Antigen Carm1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056374

 
novus biologicals canada
NB10056374
Unlabeled 0.1mg Each for $257.41

NB10056374T

 
novus biologicals canada
NB10056374SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a mixture of synthetic peptides containing amino acids 45-69 and 595-608 of human PRMT4.
Antigen Carm1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Gene Accession No. Q86X55
Gene ID (Entrez) 10498
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL