Please login to your online account to display your discounted pricing

anti-Carm1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056374SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056374T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 10498
Formulation PBS containing 0.05% BSA
Gene Accession No. Q86X55
Primary or Secondary Primary
Concentration 0.5mg/mL
Format Purified
Quantity 0.025mg
Immunogen This antibody was developed against a mixture of synthetic peptides containing amino acids 45-69 and 595-608 of human PRMT4.
Cross Reactivity Human
Applications Western Blotting
Conjugate Unlabeled
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Antigen Carm1
Host Species Rabbit