Please login to your online account to display your discounted pricing

anti-CARD6, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056110

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056110

  • / Each
 

Description & Specifications

Specifications

Format Antiserum
Formulation PBS
Gene ID (Entrez) 84674
Test Specificity CARD6 .
Cross Reactivity Human
Host Species Rabbit
Antigen CARD6
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.05mL
Immunogen A synthetic peptide corresponding to amino acids 113-131 (NSEDAFSPGIKQPEAPEIT) of human CARD6 was used as immunogen
Primary or Secondary Primary
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO