Please login to your online account to display your discounted pricing

anti-CARD6, Polyclonal, Novus Biologicals™

$144.63 - $222.95

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 113-131 (NSEDAFSPGIKQPEAPEIT) of human CARD6 was used as immunogen
Antigen CARD6
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056110

 
novus biologicals canada
NB10056110
Unlabeled 0.05mL Each for $222.95

NB10056110T

 
novus biologicals canada
NB10056110SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 113-131 (NSEDAFSPGIKQPEAPEIT) of human CARD6 was used as immunogen
Antigen CARD6
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity CARD6 .
Gene ID (Entrez) 84674
Formulation PBS