Please login to your online account to display your discounted pricing

anti-CARD6, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056111SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056111T

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Gene ID (Entrez) 84674
Test Specificity CARD6 .
Conjugate Unlabeled
Formulation PBS
Regulatory Status RUO
Host Species Rabbit
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen A synthetic peptide corresponding to amino acids 156-173 (RETALSARKNEKEYDTPE) of human CARD6 was used as immunogen; GenBank no. gi|16554564|ref|NP_115976.2|. The immunogen sequence is 100% conserved in human CINCIN1
Format Antiserum
Primary or Secondary Primary
Quantity 0.025mL
Monoclonal or Polyclonal Polyclonal
Antigen CARD6