Please login to your online account to display your discounted pricing

anti-CARD6, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056111

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056111

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 84674
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Immunogen A synthetic peptide corresponding to amino acids 156-173 (RETALSARKNEKEYDTPE) of human CARD6 was used as immunogen; GenBank no. gi|16554564|ref|NP_115976.2|. The immunogen sequence is 100% conserved in human CINCIN1
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity CARD6 .
Formulation PBS
Regulatory Status RUO
Antigen CARD6
Host Species Rabbit
Cross Reactivity Human
Conjugate Unlabeled
Quantity 0.05mL
Format Antiserum