Please login to your online account to display your discounted pricing

anti-CARD6, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056110SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056110T

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 84674
Conjugate Unlabeled
Test Specificity CARD6 .
Primary or Secondary Primary
Quantity 0.025mL
Cross Reactivity Human
Formulation PBS
Immunogen A synthetic peptide corresponding to amino acids 113-131 (NSEDAFSPGIKQPEAPEIT) of human CARD6 was used as immunogen
Antigen CARD6
Format Antiserum
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Host Species Rabbit
Regulatory Status RUO