Please login to your online account to display your discounted pricing

anti-CARD12, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP178979

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP178979

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Formulation PBS, 30% glycerol
Primary or Secondary Primary
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Gene ID (Entrez) 58484
Gene Accession No. Q9NPP4
Host Species Rabbit
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Quantity 0.1mL
Concentration 1.13mg/mL
Format Affinity Purified
Immunogen A synthetic peptide made to an internal portion of the human CARD12 protein (between residues 600-700) [UniProt Q9NPP4]
Antigen CARD12
Regulatory Status RUO