Please login to your online account to display your discounted pricing

anti-CARD12, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056143SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056143T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 58484
Formulation PBS
Conjugate Unlabeled
Test Specificity Ipaf .
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Antigen CARD12
Host Species Rabbit
Format Antiserum
Monoclonal or Polyclonal Polyclonal
Immunogen A synthetic peptide corresponding to amino acids 971-989 (DFSTKEFLPDPALVRKLSQ) of human Ipaf/Clan/CARD12 was used as immunogen; GenBank no. gi|40788015|ref|NP_067032.3|.
Quantity 0.025mL