Please login to your online account to display your discounted pricing

anti-CARD12, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056143

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056143

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 58484
Formulation PBS
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Host Species Rabbit
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Ipaf .
Quantity 0.05mL
Primary or Secondary Primary
Antigen CARD12
Cross Reactivity Human
Regulatory Status RUO
Format Antiserum
Immunogen A synthetic peptide corresponding to amino acids 971-989 (DFSTKEFLPDPALVRKLSQ) of human Ipaf/Clan/CARD12 was used as immunogen; GenBank no. gi|40788015|ref|NP_067032.3|.