Please login to your online account to display your discounted pricing

anti-CARD12, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP178980SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP178980SS

  • / Each
 

Description & Specifications

Specifications

Antigen CARD12
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Concentration 1.09mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. Q9NPP4
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human CARD12 protein (between residues 950-1015) [UniProt Q9NPP4]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 58484
Target Species Human
Target Species Murine