Please login to your online account to display your discounted pricing

anti-CARD12, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP178980

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP178980

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 58484
Conjugate Unlabeled
Regulatory Status RUO
Gene Accession No. Q9NPP4
Format Affinity Purified
Concentration 1.09mg/mL
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Formulation PBS, 30% glycerol
Cross Reactivity Human
Cross Reactivity Mouse
Quantity 0.1mL
Primary or Secondary Primary
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human CARD12 protein (between residues 950-1015) [UniProt Q9NPP4]
Monoclonal or Polyclonal Polyclonal
Antigen CARD12