Please login to your online account to display your discounted pricing

anti-CARD12, Polyclonal, Novus Biologicals™

$142.00 - $407.76

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human CARD12 protein (between residues 950-1015) [UniProt Q9NPP4]
Antigen CARD12
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP178980

 
novus biologicals canada
NBP178980
Unlabeled 0.1mL Each for $407.76

NBP178980SS

 
novus biologicals canada
NBP178980SS
Unlabeled 0.025mL Each for $142.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human CARD12 protein (between residues 950-1015) [UniProt Q9NPP4]
Antigen CARD12
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Gene Accession No. Q9NPP4
Gene ID (Entrez) 58484
Formulation PBS, 30% glycerol
Concentration 1.09mg/mL