Please login to your online account to display your discounted pricing

anti-Capicua, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to a region within residue 1150-1200 of human Capicua. [NP_055940.3]
Antigen Capicua
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11059905T

 
novus biologicals canada
NB11059905SS
Unlabeled 0.025mL Each for $109.00

NB11059905

 
novus biologicals canada
NB11059905
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to a region within residue 1150-1200 of human Capicua. [NP_055940.3]
Antigen Capicua
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Gene Accession No. Q96RK0
Gene ID (Entrez) 23152
Formulation Antigen Affinity Purified
Concentration 1mg/mL