Please login to your online account to display your discounted pricing

CAP-18 Polyclonal Antibody, Invitrogen™


CAP-18 Antibody, OSC00028G, from Invitrogen™. Species Reactivity: Human; Applications: Immunohistochemistry, Western Blot.

Manufacturer: pierce biotechnology inc  OSC00028G

Catalog No. PIOSC00028G

  • / Each
 

Description & Specifications

Specifications

Antigen CAP-18
Applications Immunohistochemistry
Applications Western Blot
Conjugate Unlabeled
Format Purified
Formulation 500μg purified antibody (1mg/mL) and no preservative
Gene Accession No. P49913
Gene Alias CAMP, CAP18, FALL39, FALL-39, 18 kDa cationic antimicrobial protein, CAP-18, hCAP-18, LL37, CRAMP, HSD26, hCAP18
Gene Symbols CAMP
Host Species Rabbit
Immunogen A synthetic peptide from human Cathelicidin antimicrobial peptide
Isotype IgG
Quantity 500μg
Storage Requirements avoiding freeze/thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 820
Target Species Human

CAP-18 Antibody, OSC00028G, from Invitrogen. Species Reactivity: Human; Applications: Immunohistochemistry, Western Blot.


Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: A synthetic peptide from human Cathelicidin antimicrobial peptide
Formulation: 500μg purified antibody (1mg/mL) and no preservative
Synonyms: CAMP, CAP18, FALL39, FALL-39, 18 kDa cationic antimicrobial protein, CAP-18, hCAP-18, LL37, CRAMP, HSD26, hCAP18
Uniprot Accession: P49913
Entrez GeneID: 820
Gene Symbol: CAMP
Reconstitute with 500 ul of distilled water.