Please login to your online account to display your discounted pricing

anti-CAP-18 Polyclonal, Thermo Scientific™

CAP-18 Polyclonal antibody OSC00039W from Thermo Scientific™ specifically detects CAP-18 in Human samples, and it is validated for Control

$448.05 - $448.05

Specifications

Antigen CAP-18
Cross Reactivity Human
Format Purified
Host Species Sheep
Immunogen A synthetic peptide from human Cathelicidin antimicrobial peptide
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

PIOSC00028G

 
pierce biotechnology inc
OSC00028G
Unlabeled 500μg Each for $448.05

PIOSC00037W

 
pierce biotechnology inc
OSC00037W
Unlabeled 150μL Each for $448.05

PIOSC00038G

 
pierce biotechnology inc
OSC00038G
Unlabeled 500μg Each for $448.05

PIOSC00039W

 
pierce biotechnology inc
OSC00039W
Unlabeled 150μL Each for $448.05
Description & Specifications

Specifications

Antigen CAP-18
Cross Reactivity Human
Format Purified
Host Species Sheep
Immunogen A synthetic peptide from human Cathelicidin antimicrobial peptide
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements avoiding freeze/thaw cycles.

CAP-18 Polyclonal antibody OSC00039W from Thermo Scientific™ specifically detects CAP-18 in Human samples, and it is validated for Control


Host Species: Rabbit
Cross Reactivity: Human
Immunogen: A synthetic scrambled peptide form of human Cathelicidin antimicrobial peptide (CAP-18, hCAP-18, antibacterial protein LL-37, CAMP, CRAMP, FALL39) conjugated to an immunogenic carrier protein has been used as the antigen
Formulation: 150μL unpurified Antibody in whole serum, with no preservative;
Synonyms: CAMP, CAP18, FALL39, FALL-39, 18 kDa cationic antimicrobial protein, CAP-18, hCAP-18, LL37, CRAMP, HSD26
Uniprot Accession: P49913
Gene Symbol: CAMP
Entrez GeneID: 820
OSC00039W detects CAP-18 in Human samples. OSC00039W has been successfully used in Control procedures. OSC00039W The immunogen is A synthetic scrambled peptide form of human Cathelicidin antimicrobial peptide (CAP-18, hCAP-18, antibacterial protein LL-37, CAMP, CRAMP, FALL39) conjugated to an immunogenic carrier protein has been used as the antigen.