Please login to your online account to display your discounted pricing

CAP-18 Polyclonal Antibody, Invitrogen™


CAP-18 Antibody, OSC00038G, from Invitrogen™. Species Reactivity: Human; Applications: Immunohistochemistry, Western Blot.

Manufacturer: pierce biotechnology inc  OSC00038G

Catalog No. PIOSC00038G

  • / Each
 

Description & Specifications

Specifications

Gene Alias CAMP, CAP18, FALL39, FALL-39, 18 kDa cationic antimicrobial protein, CAP-18, hCAP-18, LL37, CRAMP, HSD26, hCAP18
Applications Immunohistochemistry
Applications Western Blot
Formulation 500μg purified antibody (1mg/mL) and no preservative
Gene ID (Entrez) 820
Cross Reactivity Human
Storage Requirements avoiding freeze/thaw cycles.
Species Reactivity Human
Gene Accession No. P49913
Gene Symbols CAMP
Isotype IgG
Format Purified
Host Species Sheep
Quantity 500μg
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Immunogen A synthetic peptide from human Cathelicidin antimicrobial peptide
Antigen CAP-18

CAP-18 Antibody, OSC00038G, from Invitrogen. Species Reactivity: Human; Applications: Immunohistochemistry, Western Blot.


Host Species: Sheep
Cross Reactivity: Human
Isotype: IgG
Immunogen: A synthetic peptide from human Cathelicidin antimicrobial peptide
Formulation: 500μg purified antibody (1mg/mL) and no preservative
Synonyms: CAMP, CAP18, FALL39, FALL-39, 18 kDa cationic antimicrobial protein, CAP-18, hCAP-18, LL37, CRAMP, HSD26, hCAP18
Uniprot Accession: P49913
Entrez GeneID: 820
Gene Symbol: CAMP
Reconstitute with 500 ul of distilled water.