Please login to your online account to display your discounted pricing

anti-CaMKII beta, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB12022131

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB12022131

  • / Each
 

Description & Specifications

Specifications

Formulation 50% glycerol
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Gene ID (Entrez) 816
Test Specificity Human CAMKII beta GenBank no. GENE ID: 816 CAMK2B is a 664 amino acid protein. However, several isoforms of varying amino acid sizes have been described for CAMKII beta. Researchers are encouraged to utilize the sequence information used for immunogen (VRRGSGTPEAEAPRQ) to gain additional information about these isoforms and various molecular weights which may be detected by western blot analysis.
Conjugate Unlabeled
Format Antiserum
Antigen CaMKII beta
Host Species Rabbit
Regulatory Status RUO
Quantity 0.1mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Primary or Secondary Primary
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 499-513 (VRRGSGTPEAEAPRQ) of human CAMKII beta.
Monoclonal or Polyclonal Polyclonal