Please login to your online account to display your discounted pricing

anti-CaMKII beta, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 499-513 (VRRGSGTPEAEAPRQ) of human CAMKII beta.
Antigen CaMKII beta
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB12022131T

 
novus biologicals canada
NB12022131SS
Unlabeled 0.025mL Each for $139.00

NB12022131

 
novus biologicals canada
NB12022131
Unlabeled 0.1mL Each for $307.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 499-513 (VRRGSGTPEAEAPRQ) of human CAMKII beta.
Antigen CaMKII beta
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity Human CAMKII beta GenBank no. GENE ID: 816 CAMK2B is a 664 amino acid protein. However, several isoforms of varying amino acid sizes have been described for CAMKII beta. Researchers are encouraged to utilize the sequence information used for immunogen (VRRGSGTPEAEAPRQ) to gain additional information about these isoforms and various molecular weights which may be detected by western blot analysis.
Gene ID (Entrez) 816
Formulation 50% glycerol