Please login to your online account to display your discounted pricing

anti-CaMKID beta, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB12022043SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB12022043T

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 57118
Formulation PBS containing 0.2% gelatin
Quantity 0.025mg
Test Specificity CAMK 1D beta . Cross-reacts with Human. Expected to cross-react with Mouse (100% identity with immunogen) due to sequence homology. Not yet tested in other species.
Conjugate Unlabeled
Applications Western Blotting
Concentration 0.5mg/mL
Regulatory Status RUO
Antigen CaMKID beta
Cross Reactivity Human
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of human CAMK1D-beta between amino acids 10-24 (CSSWKKQAEDIKKIFE).
Primary or Secondary Primary
Format Purified
Host Species Rabbit