Please login to your online account to display your discounted pricing

anti-CaMKID beta, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB12022043

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB12022043

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Primary or Secondary Primary
Applications Western Blotting
Formulation PBS containing 0.2% gelatin
Gene ID (Entrez) 57118
Format Purified
Test Specificity CAMK 1D beta . Cross-reacts with Human. Expected to cross-react with Mouse (100% identity with immunogen) due to sequence homology. Not yet tested in other species.
Host Species Rabbit
Quantity 0.1mg
Antigen CaMKID beta
Cross Reactivity Human
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of human CAMK1D-beta between amino acids 10-24 (CSSWKKQAEDIKKIFE).
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO