Please login to your online account to display your discounted pricing

anti-CaMKID beta, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of human CAMK1D-beta between amino acids 10-24 (CSSWKKQAEDIKKIFE).
Antigen CaMKID beta
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB12022043

 
novus biologicals canada
NB12022043
Unlabeled 0.1mg Each for $255.80

NB12022043T

 
novus biologicals canada
NB12022043SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of human CAMK1D-beta between amino acids 10-24 (CSSWKKQAEDIKKIFE).
Antigen CaMKID beta
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity CAMK 1D beta . Cross-reacts with Human. Expected to cross-react with Mouse (100% identity with immunogen) due to sequence homology. Not yet tested in other species.
Gene ID (Entrez) 57118
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL