Please login to your online account to display your discounted pricing

anti-Calreticulin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600101SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600101SS

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Applications Blocking
Applications Dot Blot
Applications Electron Microscopy
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunoprecipitation
Applications Neutralization
Applications Simple Western
Applications Western Blotting
Gene ID (Entrez) 811
Quantity 0.025mL
Monoclonal or Polyclonal Polyclonal
Test Specificity Human, bovine, rat and mouse calreticulin. Others not tested.
Concentration 1.0mg/mL
Regulatory Status RUO
Antigen Calreticulin
Primary or Secondary Primary
Format Affinity Purified
Immunogen A fusion protein to mouse Calreticulin [UniProt No. P14211]
Host Species Rabbit