Please login to your online account to display your discounted pricing

anti-Calreticulin, Polyclonal, Novus Biologicals™

$144.63 - $384.00

Specifications

Host Species Rabbit
Immunogen A fusion protein to mouse Calreticulin [UniProt No. P14211]
Antigen Calreticulin
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB600101SS

 
novus biologicals canada
NB600101SS
Unlabeled 0.025mL Each for $144.63

NB600101

 
novus biologicals canada
NB600101
Unlabeled 0.1mL Each for $384.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A fusion protein to mouse Calreticulin [UniProt No. P14211]
Antigen Calreticulin
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Blocking
Applications Dot Blot
Applications Electron Microscopy
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunoprecipitation
Applications Neutralization
Applications Simple Western
Applications Western Blot
Test Specificity Human, bovine, rat and mouse calreticulin. Others not tested.
Gene ID (Entrez) 811
Concentration 1.0mg/mL