Please login to your online account to display your discounted pricing

anti-Calreticulin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600101

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600101

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Concentration 1.0mg/mL
Cross Reactivity Bovine
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Applications Blocking
Applications Dot Blot
Applications Electron Microscopy
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunoprecipitation
Applications Neutralization
Applications Simple Western
Applications Western Blotting
Conjugate Unlabeled
Gene ID (Entrez) 811
Test Specificity Human, bovine, rat and mouse calreticulin. Others not tested.
Immunogen A fusion protein to mouse Calreticulin [UniProt No. P14211]
Quantity 0.1mL
Regulatory Status RUO
Antigen Calreticulin
Host Species Rabbit
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary