Please login to your online account to display your discounted pricing

anti-Calnexin, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the canine Calnexin protein (within residues 25-100). [Swiss-Prot P24643]
Antigen Calnexin
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1001965SS

 
novus biologicals canada
NB1001965SS
Unlabeled 0.025mL Each for $139.00

NB1001965

 
novus biologicals canada
NB1001965
Unlabeled 0.1mL Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the canine Calnexin protein (within residues 25-100). [Swiss-Prot P24643]
Antigen Calnexin
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blot
Test Specificity Human, mouse, rat, Xenopus laevis, bovine, chicken, Drosophilia melanogaster, guinea pig, pig, quail, rabbit, zebrafish and sheep. Predicted to react with dog based on 100% sequence homology.
Gene ID (Entrez) 821
Formulation Tris-glycine, 150mM NaCl
Concentration 1mg/mL