missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcitonin Monoclonal Antibody (SP17), Invitrogen™
Rabbit Monoclonal Antibody
Supplier: Invitrogen MA514552
Description
MA5-14552 targets Calcitonin in IHC (P) applications and shows reactivity with Canine, Equine, Human, mouse, Non-human primate, Ovine, Porcine, and Rat samples. The MA5-14552 immunogen is synthetic human calcitonin 1-32 amino acid peptide.
Calcitonin is a 32 amino acid peptide which can be demonstrated in C cells of the normal and hyperplastic thyroid. Staining for calcitonin may be used for the identification of a spectrum of C cell proliferative abnormalities ranging from C cell hyperplasia to invasive tumors. Staining for calcitonin in medullary carcinoma of the thyroid produces a fine granular pattern in the cytoplasm. Amyloid deposits within the tumor may also exhibit varying degrees of calcitonin activity.Specifications
Calcitonin | |
Monoclonal | |
Unconjugated | |
CALCA | |
alpha-type CGRP, calcitonin, calcitonin 1, calcitonin gene-related peptide I, calcitonin/calcitonin-related polypeptide, alpha, katacalcin | |
Rabbit | |
500 μL | |
Primary | |
Canine, Equine, Human, Mouse, Non-human Primate, Ovine, Porcine, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
SP17 | |
tissue culture supernatant diluted in TBS with 1% BSA and 0.1% sodium azide; pH 7.5 | |
P41547, Q9N0T2, P01258, P70160, P01261, P30880, P01256 | |
CALCA | |
Synthetic human calcitonin 1-32 amino acid peptide | |
RUO | |
100033906, 100294596, 100579174, 12310, 24241, 403946, 796 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |