Please login to your online account to display your discounted pricing

anti-Calcitonin Clone: SP17

Calcitonin Monoclonal Ab MA5-14552 from Thermo Scientific™ specifically detects Calcitonin in Ca, Equine, Hu, Ms, Nhp, Ov, Po and Rt samples, and it is validated for IHC(P)

$578.55 - $578.55

Specifications

Antigen Calcitonin
Clone SP17
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

PIMA190591

 
pierce biotechnology inc
MA190591
Unlabeled 6mL Each for $578.55
Description & Specifications

Specifications

Antigen Calcitonin
Clone SP17
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-human primate
Cross Reactivity Ovine
Cross Reactivity Porcine
Cross Reactivity Rat
Format Purified
Host Species Rabbit
Regulatory Status RUO
Immunogen Synthetic human calcitonin 1-32 amino acid peptide.
Isotype IgG
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4° C

Calcitonin Monoclonal Ab MA5-14552 from Thermo Scientific™ specifically detects Calcitonin in Ca, Equine, Hu, Ms, Nhp, Ov, Po and Rt samples, and it is validated for IHC(P)

Calcitonin is a 32 amino acid peptide which can be demonstrated in C cells of the normal and hyperplastic thyroid. Staining for calcitonin may be used for the identification of a spectrum of C cell proliferative abnormalities ranging from C cell hyperplasia to invasive tumors. Staining for calcitonin in medullary carcinoma of the thyroid produces a fine granular pattern in the cytoplasm. Amyloid deposits within the tumor may also exhibit varying degrees of calcitonin activity.
Host Species: Rabbit
Cross Reactivity: Canine, Equine, Human, Mouse, Non-human primate, Ovine, Porcine, Rat
Isotype: IgG
Clone: SP17
Immunogen: Synthetic human calcitonin 1-32 amino acid peptide
Formulation: 500μL unpurified Antibody in tissue culture supernatant, with 15mM sodium azide;
Synonyms: alpha-type CGRP, calcitonin, calcitonin 1, calcitonin gene-related peptide I, calcitonin/calcitonin-related polypeptide, alpha, katacalcin
Uniprot Accession: P01256, P01258, P41547, Q99JA0, Q9N0V5
Gene Symbol: CALCA
Entrez GeneID: 100033906, 100294596, 100579174, 12310, 24241, 403946, 796
MA5-14552 detects Calcitonin in Canine, Equine, Human, Mouse, Non-human primate, Ovine, Porcine and Rat samples. MA5-14552 has been successfully used in Immunohistochemistry (Paraffin) procedures. MA5-14552 The immunogen is Synthetic human calcitonin 1-32 amino acid peptide.