Please login to your online account to display your discounted pricing

anti-Cactin, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 460-510 of human Cactin was used as the immunogen.
Antigen Cactin
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP170775SS

 
novus biologicals canada
NBP170775SS
Unlabeled 0.025mg Each for $139.00

NBP170775

 
novus biologicals canada
NBP170775
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 460-510 of human Cactin was used as the immunogen.
Antigen Cactin
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Cross reacts with Chimpanzee, Cow, Dog, Human, Opossum, Rhesus Monkey and Wild Pig. Predicted to react with Bacteria, Mosquito, Mouse, Pufferfish, Rat, Wild Turkey, Zebrafish. The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, rhesus monkey, 88% homologous in wild pig, 82% homologous in dog, cow, opossum, 64% homologous in mouse, rat, wild turkey, mosquito and 47% homologous in zebrafish, pufferfish, and bacteria.
Gene Accession No. Q8WUQ7
Gene ID (Entrez) 58509
Formulation PBS containing 0.05% BSA