Please login to your online account to display your discounted pricing

anti-C9RANT, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225018

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225018

  • / Each
 

Description & Specifications

Specifications

Antigen C9RANT
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity All
Format Antisera
Host Species Rabbit
Immunogen (GP)8 and (GR)8 peptides
Quantity 0.05mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Test Specificity This C9RANT antibody detects poly(GP) peptides produced via repeat associated non-ATG translation in central nervous system tissues from humans affected by an expanded hexanucleotide (GGGGCC) repeat in the C9ORF72 gene, the most common genetic cause of amyotrophic lateral sclerosis and frontotemporal dementia (now termed c9FTD/ALS). All species in which poly(GP) peptides are synthesized
Target Species All