Please login to your online account to display your discounted pricing

anti-C9RANT, Polyclonal, Novus Biologicals™

$214.00 - $329.00

Specifications

Host Species Rabbit
Immunogen (GP)8 and (GR)8 peptides
Antigen C9RANT
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP225018SS

 
novus biologicals canada
NBP225018SS
Unlabeled 0.025mL Each for $214.00

NBP225018

 
novus biologicals canada
NBP225018
Unlabeled 0.05mL Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen (GP)8 and (GR)8 peptides
Antigen C9RANT
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity This C9RANT antibody detects poly(GP) peptides produced via repeat associated non-ATG translation in central nervous system tissues from humans affected by an expanded hexanucleotide (GGGGCC) repeat in the C9ORF72 gene, the most common genetic cause of amyotrophic lateral sclerosis and frontotemporal dementia (now termed c9FTD/ALS). All species in which poly(GP) peptides are synthesized