Please login to your online account to display your discounted pricing

anti-C5L2/GPR77, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056497

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056497

  • / Each
 

Description & Specifications

Specifications

Formulation PBS containing 0.2% gelatin
Monoclonal or Polyclonal Polyclonal
Gene Accession No. Q9P296
Conjugate Unlabeled
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, and chimp (isoforms 1, 2, 3, and 4), 88% homologous in rhesus monkey (isoforms 1, 2, 3), and 76% homologous in bovine (isoforms 1 and 2). Cross reacts with Chimpanzee and Human. Predicted to react with Bovine and Rhesus Monkey.
Regulatory Status RUO
Gene ID (Entrez) 27202
Antigen C5L2/GPR77
Host Species Rabbit
Cross Reactivity Human
Cross Reactivity Primate
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Format Affinity Purified
Immunogen A portion of amino acids 1-50 of human GPR77 was used as the immunogen.
Primary or Secondary Primary
Quantity 0.1mg