Please login to your online account to display your discounted pricing

anti-C5L2/GPR77, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human GPR77 was used as the immunogen.
Antigen C5L2/GPR77
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056497

 
novus biologicals canada
NB10056497
Unlabeled 0.1mg Each for $307.00

NB10056497T

 
novus biologicals canada
NB10056497SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human GPR77 was used as the immunogen.
Antigen C5L2/GPR77
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, and chimp (isoforms 1, 2, 3, and 4), 88% homologous in rhesus monkey (isoforms 1, 2, 3), and 76% homologous in bovine (isoforms 1 and 2). Cross reacts with Chimpanzee and Human. Predicted to react with Bovine and Rhesus Monkey.
Gene Accession No. Q9P296
Gene ID (Entrez) 27202
Formulation PBS containing 0.2% gelatin