Please login to your online account to display your discounted pricing

anti-C5L2/GPR77, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056497SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056497T

  • / Each
 

Description & Specifications

Specifications

Antigen C5L2/GPR77
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Format Affinity Purified
Formulation PBS containing 0.2% gelatin
Gene Accession No. Q9P296
Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human GPR77 was used as the immunogen.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 27202
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, and chimp (isoforms 1, 2, 3, and 4), 88% homologous in rhesus monkey (isoforms 1, 2, 3), and 76% homologous in bovine (isoforms 1 and 2). Cross reacts with Chimpanzee and Human. Predicted to react with Bovine and Rhesus Monkey.
Target Species Human
Target Species Primate